Cyclohexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexahydro-Benzene, Hexahydrobenzene, Hexamethylene
M-Clarity™ quality level = MQ100
1.02827.1000
1.02827.2500
EM1.02827.1000CS
1318
USD
EM1.02827.1000
EM1.02827.2500
Cyclohexane, LiChrosolv® for liquid chromatography, Supelco®
Cyclohexane
Formula:
C₆H₁₂ Boiling Pt: 81 °C (1013 hPa) Melting Pt: 7 °C Density: 0.7785 g/cm³ (20 °C) Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 13,02752 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...