Pyridine, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: AzabenzeneAzine

M-Clarity™ quality level = MQ200

1.07462.6026 1.07462.6026 1.07462.1000 1.07462.9190 1.07462.1000 1.07462.9190 1.07462.2500 1.07462.2500
EM1.07462.1000CS 1222 USD
CA1.07462.2500 EM1.07462.1000 CA1.07462.9190 EM1.07462.6026 EM1.07462.2500 CA1.07462.6026 EM1.07462.9190 CA1.07462.1000
Pyridine, EMPLURA®, Supelco®
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.982 g/cm3 at 20 °C
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 12,08153
https://us.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR