Boric acid 99.5-100.5% (by alkalimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

M-Clarity™ quality level = MQ300

1.00165.9025 1.00165.9025
EM1.00165.9025EA 2086 USD
CA1.00165.9025 EM1.00165.9025
Boric acid 99.5-100.5% (by alkalimetry), EMSURE® for analysis, Supelco®
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C (dec., lit.)
Density: 1.489 g/cm3 (23 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://us.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR