Cyclohexanone ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pimelic Ketone

M-Clarity™ quality level = MQ200

1.02888.1000 1.02888.2500 1.02888.2500 1.02888.6010 1.02888.9191 1.02888.6025 1.02888.6025 1.02888.9191 1.02888.6010 1.02888.1000
EM1.02888.9191EA 6190.3 USD
EM1.02888.9191 EM1.02888.6025 EM1.02888.1000 CA1.02888.6010 EM1.02888.2500 CA1.02888.6025 CA1.02888.1000 EM1.02888.6010 CA1.02888.9191 CA1.02888.2500
Cyclohexanone ≥99% (by GC), EMPLURA®, Supelco®
Cyclohexanone
Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.95 g/cm3 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://us.vwr.com/stibo/web/std.lang.all/12/57/771257.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR