Toluene ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

M-Clarity™ quality level = MQ200

1.08323.1000 1.08323.2500 1.08323.6025 1.08323.9011 1.08323.9190
EM1.08323.1000CS 415 USD
EM1.08323.1000 EM1.08323.2500 EM1.08323.6025 EM1.08323.9011 EM1.08323.9190
Toluene ≥99% (by GC), EMPLURA®, Supelco®
Toluene

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 (lit.)
Melting Pt: -95 (lit.)
Density: 0.865 g/ml (25°C) (lit.)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR