Formic acid 98-100%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methanoic acid
, Aminic Acid
M-Clarity™ quality level = MQ100
1.11670.1000
EM1.11670.1000CS
4674
USD
EM1.11670.1000
Formic acid 98-100%, Suprapur® for trace metal analysis, Supelco®
Formic acid
Formula:
HCO₂H MW: 46.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8 (3), |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...