Ethyl acetate, EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.22277.2500
8.22277.5000
EM8.22277.2500CS
388
USD
EM8.22277.2500
EM8.22277.5000
Ethyl acetate, EMPLURA®, Supelco®
Ethyl acetate
Formula:
CH₃COOC₂H₅ Boiling Pt: 77.1 °C (1013 hPa) Melting Pt: –83 °C Density: 0.902 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...