Silica gel, with moisture indicator (brown gel), Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.01972.1000 1.01972.5000 1.01972.9025
EM1.01972.1000CS 740 USD
EM1.01972.1000 EM1.01972.5000 EM1.01972.9025
Silica gel, with moisture indicator (brown gel), Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR