L-Threonine ≥98.5%, Ultrapure
Supplier: Thermo Fisher Scientific
Synonyms:
(2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
J22290-22
J22290-A1
101171-176EA
144.55
USD
101171-176
101176-060
L-Threonine ≥98.5%, Ultrapure
L-Threonine
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064270 CAS Number: 72-19-5 EINECS: 200-774-1 Merck Index: 14,9380 |
Specification Test Results
Assay | ≥98.5% |
Impurity Content Sulphate (SO₄) | ≤0.03% |
Loss on Drying | ≤0.2% |
Optical Rotation | –26.7 to –29.1° (c=6 in H₂O) |
pH (5%, H₂O) | 5.0 to 6.5 |
Identification (FTIR) | Conforms |
Chloride Content | ≤0.05% |
Residue on Ignition | ≤0.4% |
Heavy Metals (Pb) | ≤15 ppm |
Iron (Fe) | ≤30 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...