L(-)-Tryptophan 98.5-101.5% (dry basis)

Supplier: Thermo Fisher Scientific

Synonyms: L-α-Amino-3-indolepropionic acid(S)-2-Amino-3-(3-indolyl)propionic acidH-Trp-OHL-TryptophanTryptophan

J22765-A1 J22765-22
101170-168EA 129.6 USD
101170-168 101174-006
L(-)-Tryptophan 98.5-101.5% (dry basis)
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 280…285 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 14,09797

Order Now

Specification Test Results

Assay (Dry Basis) 98.5 to 101.5%
Chloride Content ≤0.05%
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content, Organic Impurities Meets the Requirements
Impurity Content Sulphate (SO₄) ≤0.03%
Loss on Drying ≤0.3%
Optical Rotation –29.4 to –32.8° (c=1 in H₂O)
pH (1%, H₂O) 5.5 to 7.0
Heavy Metals (Pb) ≤15 ppm
Iron (Fe) ≤0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR