Dextran sulfate sodium salt, M.W. 40000 - 50000
Supplier: Thermo Fisher Scientific
Synonyms:
Dextran sulfate sodium from Dextran
, Dextran sulphate sodium from Dextran
J14489-09
J14489-18
J14489-22
101175-484EA
599
USD
101175-484
101170-838
101175-006
Dextran sulfate sodium salt, M.W. 40000 - 50000
Dextran sulphate sodium salt
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Ambient |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...