Citric acid, anhydrous 99.5-100.05% (dry basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
J13729-36
J13729-A1
101174-286EA
76.89
USD
101174-286
101173-550
Citric acid, anhydrous 99.5-100.05% (dry basis)
Citric acid
Formula:
HOC(COOH)(CH₂COOH)₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 153…154 °C Density: 1.665 Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 14,2326 |
|
Specification Test Results
Assay | 99.5 - 100.5% (dry basis) |
Assay, Readily Carbonizable Substances | Passes Test |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.1% |
Impurity Content, Oxalate | Passes Test |
Impurity Content, Sulphate (SO₄) | Passes test |
Water Content (Karl Fischer Titration) | ≤1.0% |
Heavy Metals (Pb) | ≤10 µg/gm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...