Dithiothreitol (DTT, Cleland's reagent) ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: DL-1,4-DithiothreitolDTTcis-2,3-Dihydroxy-1,4-butanedithiol(R*,R*)-1,4-Dimercapto-2,3-butandiol

J15395-22
101175-932EA 1430.58 USD
101175-932
Dithiothreitol (DTT, Cleland's reagent) ≥97%
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 125…130 °C/12 mm
Melting Pt: 40…43 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
Merck Index: 14,3376
https://us.vwr.com/stibo/web/std.lang.all/98/19/2299819.jpg

Order Now

Specification Test Results

Assay (unspecified) ≥97%
Identification (FTIR) Conforms
Impurity Content, Oxidised DTT ≤1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR