EDTA disodium salt dihydrate 0.5 M in aqueous solution, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Warning

J15694-AP J15694-AE
101170-870EA 103.8 USD
101170-870 101172-488
EDTA disodium salt dihydrate 0.5 M in aqueous solution, Ultrapure for molecular biology
EDTA disodium salt dihydrate

0.5 M solution of EDTA.Na₂ in high purity dH₂O, pH 8.0.


Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.


Packaging: Dispensed into durable, square Nalgene bottles.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR