Cesium chloride 99% (metals basis), Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cesium Chloride

J75822-A1 J75822-36
101171-412EA 845.62 USD
101171-412 101175-326 101174-850
Cesium chloride 99% (metals basis), Ultrapure for molecular biology
Cesium chloride
Formula: CsCl
Boiling Pt: 1303 °C
Melting Pt: 646 °C
Density: 3.99
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 14,2011

Order Now

Specification Test Results

Assay (Unspecified) 100.00%
Assay (Unspecified), Absorbance (260 nm, 50% w/v, 1 cm) ≤0.02
Dnase, (endo) Passes Test
Dnase, (exo) Passes Test
Impurity Content, Sulfate (SO₄) ≤5 ppm
Insoluble Material Negligible
Refractive Index, (50% w/w, 20 °C) 1.389 - 1.391
RNase Passes Test
Aluminum (Al) ≤1 ppm
Barium (Ba) ≤2 ppm
Calcium (Ca) ≤3 ppm
Chromium (Cr) ≤1 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤2 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR