Taurine ≥98.5%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific

Synonyms: 2-Aminoethanesulphonic acid2-Aminoethanesulfonic acid

J75824-A1
101174-114EA 296.58 USD
101174-114
Taurine ≥98.5%, Ultrapure for molecular biology
Taurine
Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 14,9074

Order Now

Specification Test Results

Assay (unspecified) Absorbance (260 nm, 0.5 M, 1 cm) ≤0.2
Assay (unspecified) Absorbance (280 nm, 0.5 M, 1 cm) ≤0.2
Assay (unspecified) Ammonia (NH₃) ≤0.02%
Assay (unspecified) Use Test Functionally Tested for Electrophoresis
Assay (unspecified) ≥98.5%
Chloride Content ≤0.01%
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulfate (SO₄) ≤0.01%
pH (5%, H₂O) 4.0 to 7.0
Solution test (0.5 M, H₂O) Clear and Colorless
Arsenic (As) ≤2 ppm
Heavy Metals ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR