Sodium chloride ≥99.0% ACS, Ultrapure
Supplier: Thermo Fisher Scientific
Synonyms:
Saline, NaCl, Sea salt, Halite
J21618-A1
J21618-36
J21618-A9
J21618-A7
101171-820EA
102
USD
101171-820
101172-982
101175-162
101171-822
Sodium chloride ≥99.0% ACS, Ultrapure
Sodium chloride
Formula:
NaCl Boiling Pt: 1413 °C Melting Pt: 801 °C Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Specification Test Results
Assay Chlorate and Nitrate (NO₃) | ≤0.003% |
Assay | ≥99.0% |
Residual Halide Bromide (Br) | ≤0.01% |
Residual Halide Iodide (I) | ≤0.002% |
Impurity Content Phosphate (PO₄) | ≤5 ppm |
Impurity Content Sulfate (SO₄) | ≤0.004% |
Insoluble Material | ≤0.005% |
pH (5%, H₂O, 25 °C) | 5.0 - 9.0 |
Barium (Ba) | Passes Test |
Calcium (Ca) | ≤0.002% |
Heavy Metal (Pb) | ≤5 ppm |
Iron (Fe) | ≤2 ppm |
Magnesium (Mg) | ≤0.001% |
Potassium (K) | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...