Screw Cap Micro Tubes, Type D, Sarstedt
Supplier: Sarstedt
72.607 72.692 72.692.105 72.687 72.692.100 72.692.005
101093-577PK 111.31 USD
101093-577 101093-610 101093-869 101093-748 101433-358 101093-785
Screw Cap Micro Tubes, Type D, Sarstedt
Tubes Microtubes
Type D micro tubes are designed for transport, storage, and sample processing in the laboratory
All tubes feature a non-skirted base, and are available with or without attached screw cap. Tubes and caps are manufactured from polypropylene, and are autoclavable at 121 °C. Knurls enable single-handed operation. A neutral loop screw cap can be provided with inserts for color coding.