Riser Shelf, ESD, for RTW Table, Production Basics
Supplier: Production Basics
8472 8473 8471
11005-338EA 517.65 USD
11005-338 11005-340 11005-336
Riser Shelf, ESD, for RTW Table, Production Basics
Furniture Shelves
Mounts to the top of the RTW table to provide extra workspace and privacy for operator.
  • Metal shelf frame with ESD laminate surface
  • Thickness: 3.2 cm (1.25")
Measures within static dissipative range of 10⁶ to 10⁹. Finished with rounded corners and protective T-mold edge.
For use with any Production Basics RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench.
Includes 1 ESD laminate shelf, 1 riser frame, 1 ground bolt and wire, 1 parts and hardware packet.
Ordering information: When purchased at same time as RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench the colors will be made to match. If purchasing at separate time, specify riser frame and laminate colors otherwise will be default blue frame/gray ESD laminate. Frame available in light neutral/almond, blue, white, gray and black. Worksurface available in white, gray and beige. Accessory is ready to mount on a RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench of same width.