C-Leg Quad Stand-Alone Laboratory Benches, ESD, Production Basics
Supplier: Production Basics
1340 1351 1343 1345 1346 1350 1341
11005-062EA 4412.49 USD
11005-062 11005-072 11005-064 11005-066 11005-068 11005-070 87000-246
C-Leg Quad Stand-Alone Laboratory Benches, ESD, Production Basics
Furniture Tables and Workbenches
This package includes ESD worksurfaces, necessary components and hardware to construct a four-station configuration.
  • Ideal for laboratory research and testing cells
  • Measures within static dissipative range of 10⁶ - 10⁹
Finished with rounded corners and protective T-mold edge.
Ordering information: Specify a Frame and Laminate color otherwise will be default Blue Frame/Grey ESD Laminate. Frame available in Light Neutral/Almond, Blue, White, Grey and Black. Worksurface available in White, Grey and Beige. The two (2) included 162.6 cm (64") high integrated double-sided back frames are ready to accept accessories. Accessories are sold separately. Order same width accessory as workstation width.
Delivery information: Includes (2) double-sided frame, (4) C-leg, adjustable, (2) 2C-leg, adjustable, (4) ESD worksurface, (8) support rail, (4) ground bolt and wire, (16) leveling feet, (1) parts and hardware packet.