Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]

Supplier: Tonbo Biosciences
50-0861-U025 50-0861-U100
10051-278EA 51.7 USD
10051-278 10051-280
Anti-CD86 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PO3.1]
Antibodies
The PO3.1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response.

The PO3.1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: PO3.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR