Anti-CD90.2 (30-H12) Rat Monoclonal Antibody [clone: 30-H12] (FITC)

Supplier: Tonbo Biosciences
35-0903-U025 35-0903-U100
10050-954EA 22 USD
10050-954 10050-956
Anti-CD90.2 (30-H12) Rat Monoclonal Antibody [clone: 30-H12] (FITC)
Antibodies
The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.

Type: Primary
Antigen: CD90.2
Clonality:
Clone: 30-H12
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR