GeneMate Autoclavable Waste Disposal Bags, MTC Bio
Supplier: GeneMate
490016-622 490016-624 490016-626 490016-628 490016-630
490016-622CS 221.22 USD
490016-622 490016-624 490016-626 490016-628 490016-630
GeneMate Autoclavable Waste Disposal Bags, MTC Bio
Bags Waste Disposal Bags
These high temperature autoclavable bags are manufactured from heavy gauge polypropylene material that is not only more puncture-resistant than polyethylene, but also withstands autoclaving at temperatures up to 136 °C (250 °F).
  • Each bag has an oversized white marking panel
  • Suitable for use with all types of permanent markers
Designed to resist stretching and poking, these bags are extremely pliable, leak-resistant and have seamless bottoms for exceptional strength and integrity.
Their ability to contain is completely unaffected at standard autoclave temperature (121 °C).
Certifications: They are also prominently marked with the words 'Hazardous Waste' and the standard 'Biohazard' symbol, conforming to the OSHA standard.