Pyridine-Barbituric acid APHA Cyanide_determination

Supplier: Ricca Chemical
Danger

6584-16 6584-32
RC6584-16EA 167 USD
RC6584-16 RC6584-32
Pyridine-Barbituric acid APHA Cyanide_determination
Pyridine-Barbituric acid

This reagent contains 6% (w/v) Reagent Grade Barbituric Acid, CAS No. 67-52-7, 30% (v/v) ACS Reagent Grade Pyridine, CAS No. 110-86-1, and 6% (v/v) ACS Reagent Grade Hydrochloric Acid, CAS No. 7647-01-0, in ACS Reagent Grade Water, CAS No. 7732-18-5.

Storage Temperature: Refrigerator
UN: 2924
ADR: 3 (8) III

Order Now

Specification Test Results

Appearance Golden yellow liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR