Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica, Ricca Chemical Company
Supplier: Ricca Chemical
609.2-16
609.2-4
RC609.2-16EA
68
USD
RC609.2-16
RC609.2-4
Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica, Ricca Chemical Company
Environmental Test Kits
Suitable for use in the following compendial methods:
-APHA 4500-SiO2 D, Reagent 3
-ASTM D859, Reagent 8.3