Acetic acid 1.00 N

Supplier: Ricca Chemical
Danger

Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

150-16
RC150-16EA 33.39 USD
RC150-16
Acetic acid 1.00 N
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0084 g/cm³
Flash Pt: 39 °C (AcOH)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://us.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Specification Test Results

Assay 1 ± 0.002 N at 20 °C
NIST SRM 84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR