Types Of Insect Legs
Supplier: ED SPELDY EAST CO. LTD.
T215
470150-968EA 43.25 USD
470150-968
Types Of Insect Legs
Specimens Preserved Specimens Display Specimens
This kit studies the types of insect legs
There is 1 specimen block in this package (5.5x2.5x 1in.). Specimens are preserved and encased in clear resin material. They can be observed from every side to see details. Specimens include: Fossorial leg (Mole cricket, Grylltalpa orientalis), Cursorial leg (Beetle, Cincindela sylvatica), Raptorial leg (Praying mantis, Hierodula petellifera), Saltatorial leg (Grasshopper, Aularches miliaris), Natatorial leg (Great Diving beetle, Dytiscus marginalis), and Corbicula leg (Honey bee, Apis cerana).Teacher information guide and student activity sheet are included. Size of product is 8.25x4.75x1.5in.