EZ-PECT™, Microbiologics
Supplier: Microbiologics
EZ-PECT™
0485-PEC 0484-PEC 0443-PEC 0486-PEC 0483-PEC 0581-PEC 0392-PEC
10142-718EA 861.77 USD
10142-718 10142-714 10142-736 10142-716 10142-720 10142-722 10142-712
EZ-PECT™, Microbiologics
Controls and Calibration Verification Materials
Antimicrobial Effectiveness and Preservative Efficacy Testing are a critical element to the development of safe and effective Cosmetic and Pharmaceutical products
EZ-PEC™ kits include 20 lyophilized pellets of a single enumerated QC microorganism, 10 vials of hydrating fluid (2 mL per vial) and Peel-Off Certificate of Assay. Quantitative testing saves time and reduces labor by delivering a specific number of colony forming units (CFU).
Ready-to-use EZ-PEC™ quantitative QC microorganism preparations are designed to provide the required microorganism challenge concentrations for Antimicrobial Effectiveness Tests and Preservative Efficacy Tests. When processed as directed, EZ-PEC™ delivers a final concentration of 1.0x105 to 1.0x106 CFU per mL of the product being tested.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.