PTFE Tube Compression Reducing Union, Ace Glass Incorporated
Supplier: Ace Glass
12711-02 12711-04 12711-06 12711-08 12711-12 12711-14 12711-16 12711-18 12711-20
16001-984PK 240.88 USD
16001-984 10099-318 10099-320 10099-322 10099-324 10099-326 10099-328 10099-330 10099-332 10099-334 10099-336
PTFE Tube Compression Reducing Union, Ace Glass Incorporated
Fittings and Connectors
PTFE Tube Compression Reducing Union
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions is 120 PSI for 1/16" fittings linear decreasing to 80 PSI for 3/4" fittings and 60 PSI for fittings larger than 3/4". Performance at elevated temperatures up to 85°C/185°F: 90 PSI for 1/16" fittings linear decreasing to 60 PSI for 3/4" fittings and 40 PSI for fittings larger than 3/4".