Prominence HPLC/UFLC/UFLCXR, Shimadzu
Supplier: Shimadzu
228-45136-42 228-45211-58 228-45150-42 228-45093-93 228-45203-41 228-45121-32 228-45040-58 228-45137-42 228-45089-92
10122-468EA 7536.36 USD
10122-468 10122-510 10122-488 10122-516 10122-348 10122-502 10122-508 10122-360 10122-492
Prominence HPLC/UFLC/UFLCXR, Shimadzu
Autosamplers
The modular Prominence HPLC series delivers even more accurate and reproducible data while maintaining the flexibility for which Shimadzu HPLC instrumentation is known.
Capable of system pressures in excess of 9500 psi (66MPa), and accompanied by the XR lineup of columns that features a choice of seven bonded phases, the Prominence UFLCXR delivers powerful high-pressure analysis–without sacrificing performance, reliability, or ruggedness. Utilizing standard components, Prominence UFLC raises total system productivity by generating high-quality, reproducible data without compromise.
Key components include SIL-20A/AC autosamplers, which deliver virtually zero carryover and a cycle time of just 10 seconds, LC-20AD/AB/AT Pumps, which provide unmatched gradient speed and precision, CBM-20A/Alite–The world’s first web-based controller, the CBM enables users to setup, control, and monitor their HPLC remotely; the speed of these controllers makes them ideal for ultrafast LC, SPD-20A/AV/M20A UV/PDA Detectors, which ensure fast sampling with exceptional sensitivity and stability, RF-20A/Axs Fluorescence Detectors–The highest sensitivity levels, to meet the demanding analysis requirements of trace-level components while retaining the acquisition speeds necessary for ultrafast analysis; other detector options include ELSD, RID, and CDD.
The modular components interact with the precision of an integrated system while allowing innumerable configurations–letting you design the system that best fits your needs.
Certifications: UL Listed and CSA Listed.
Ordering information: To order, contact your Avantor sales representative