MES sodium salt, Reagent Grade

Supplier: VWR

Synonyms: Sodium 2-MorpholinoethanesulfonateSodium 2-morpholinoethane-1-sulfonateMES-NaSodium 3-morpholin-1-ylethylsulfonate2-(N-Morpholino)ethanesulphonic acid sodium saltSodium 3-morpholin-1-ylethylsulphonate2-(N-Morpholino)ethanesulfonic acid sodium saltSodium 2-morpholinoethane-1-sulphonateSodium 2-Morpholinoethanesulphonate

VWRVX218-500G
89399-838EA 655.04 USD
89399-838
MES sodium salt, Reagent Grade
MES sodium salt
Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2

Order Now

Specification Test Results

Appearance White crystalline powder
Moisture = 10 %
pH (1% Water) @ 25°C 9 - 10.5
Purity = 98 %
Solubility (10%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR