Ethyl acetate, Reagent Grade
Supplier: VWR
|
Danger
|
Synonyms:
EtOAc
, Acetidin
, Ethyl ethanoate
, Vinegar naphtha
, Ethyl acetic ester
, Acetic acid ethyl ester
, Acetic ether
, Acetoxyethane
VWRVN774-1L
89422-816CS
530.89
USD
89422-816
Ethyl acetate, Reagent Grade
Ethyl acetate
Formula:
CH₃COOC₂H₅ MW: 88.11 g/mol Boiling Pt: 77.1 °C (1013 hPa) Melting Pt: –83 °C Density: 0.902 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 13,03792 |
Specification Test Results
Identification | PASS |
Purity (GC) |
|
Specific Gravity @25C | 0.894 - 0.898 |
Water (KF) |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...