GeneMate 96-Well PCR Plates, Semi-Skirted, ABI-Shaped Raised Rims, Scientific Specialties
Supplier: GeneMate
490003-822 490003-826
490003-822BX 116.58 USD
490003-822 490003-826
GeneMate 96-Well PCR Plates, Semi-Skirted, ABI-Shaped Raised Rims, Scientific Specialties
Microplates PCR Plates
The raised upper ridge on plates is designed to fit ABI-instruments using standard (0.2mL) PCR plates. Plates can be sealed with strip caps, sealing mats, or sealing film.
  • ABI-Style
  • Compatible with most commercially available thermal cyclers
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Contrasting printed alphanumeric indexing
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • High contrast black alphanumerics for rapid sample identification
  • Crystal-clear clarity
  • Easy to visualize content of each well
  • Certified free from DNase, RNase, and detectable DNA
Ordering information: Plates are available either pre-barcoded or customer-barcoded for convenient sample identification and tracking. Also available in a full range of colors. Contact your sales representative for more information.