Anti-IL12B Polyclonal Antibody

Supplier: Diagnostic Biosystems

PDR055 RP163-05
10065-016EA 368.45 USD
10065-016 10065-174
Anti-IL12B Polyclonal Antibody
Antibodies
p40 is a relatively unknown antibody that recognizes ΔNp63-a p63 isoform suggested to be highly specific for squamous/basal cells. In a recent study, p40 is equivalent to p63 in sensitivity for squamous cell carcinoma, but it is markedly superior to p63 in specificity1, which eliminates a potential pitfall of misinterpreting a p63-positive adenocarcinoma or unsuspected lymphoma as squamous cell carcinoma. These findings strongly support the routine use of p40 in place of p63 for the diagnosis of pulmonary squamous cell carcinoma.

Type:
Antigen: IL12B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR