UnoVue™ Rabbit AP Detection Reagent, Diagnostic BioSystems
Supplier: Diagnostic Biosystems
RU-AP100 RU-AP1000
10065-152EA 374.73 USD
10065-152 10065-154
UnoVue™ Rabbit AP Detection Reagent, Diagnostic BioSystems
Assays Immunological Assays
UnoVue Rabbit AP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections.
The UnoVue detection reagent may also be used with blood smears, cytosmears, and cell preparations.