UnoVue™ Mouse/Rabbit HRP Detection Reagent, Detection Systems, Diagnostic BioSystems
Supplier: Diagnostic Biosystems.jpg)
MRU-HRP100
MRU-HRP1000
10065-132EA
408.83
USD
10065-132
10065-134
UnoVue™ Mouse/Rabbit HRP Detection Reagent, Detection Systems, Diagnostic BioSystems
Assays
Immunological Assays
UnoVue Mouse/Rabbit HRP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections.
The UnoVue detection reagent may also be used with blood smears, cytosmears, and cell preparations.