Anti-CD1A Mouse Monoclonal Antibody [clone: O10]

Supplier: Diagnostic BioSystems

PDM173 MOB363 MOB363-05
10065-066EA 563.98 USD
10065-066 99990-678 10064-998
Anti-CD1A Mouse Monoclonal Antibody [clone: O10]
Antibodies
This antibody reacts with a 49 kDa protein. At least five CD1 genes are identified (CD1a, b, c, d and e). This antibody reacts with cortical thymocytes, Langerhans cells in epidermis, dendritic cells of dermis and Langerhans cells of mucosa of tonsil. This antibody may be useful in the characterization of leukemias and lymphomas.

Type:
Antigen: CD1A
Clonality: Monoclonal
Clone: O10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR