Anti-HSP90AB1 Rat Monoclonal Antibody [clone: 9G10]

Supplier: Rockland Immunochemicals
200-501-F39
RL200-501-F39EA 532 USD
RL200-501-F39
Anti-HSP90AB1 Rat Monoclonal Antibody [clone: 9G10]
Antibodies
Anti-Grp94 Antibody is suitable for WB, IP, and Flow cytometry. Expect a band approximately 98kDa Protein corresponding to the molecular mass of Grp94 on SDS PAGE immunoblots. This monoclonal antibody reacts with purified Grp94.

Recommended Dilutions: Western Blot: 0.5µg/ml;

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Guinea Pig, Hamster, Horse, Monkey, Pig, Rabbit, Sheep, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR