Anti-HSPD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-F62
RL200-401-F62EA 501 USD
RL200-401-F62
Anti-HSPD1 Rabbit Polyclonal Antibody
Antibodies
Anti-Hsp60 Antibody is suitable for use in WB, ELISA, Immunochemistryand IP. Expect a band approximately ~60kDa Protein corresponding to the molecular mass of Hsp60 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000;

Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR