Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C12-H11]

Supplier: Rockland Immunochemicals
200-301-F53
RL200-301-F53EA 532 USD
RL200-301-F53
Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C12-H11]
Antibodies
Anti-Hsp22 Antibody is suitable for use inWB, Immunochemistryand ELISA. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: 3C12-H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR