Anti-IRF5 Mouse Monoclonal Antibody [clone: 11F4A09]

Supplier: Biolegend
652601 652602
652601-BLEA 165.5 USD
652601-BL 652602-BL
Anti-IRF5 Mouse Monoclonal Antibody [clone: 11F4A09]
Antibodies
Purified anti-IRF5 [11F4A09]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5-1.0 μg per ml. For immunoprecipitation, the suggested use of this reagent is 2-10 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IRF5
Clonality: Monoclonal
Clone: 11F4A09
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR