Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RTK4530]

Supplier: Biolegend
400645 400646
400645-BLEA 213.97 USD
400645-BL 10761-186
Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: RTK4530]
Antibodies
Brilliant Violet 510™ Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK4530
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR