Anti-Galectin-9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9M1-3]

Supplier: Biolegend
348907 348908
348907-BLEA 207.2 USD
348907-BL 348908-BL
Anti-Galectin-9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9M1-3]
Antibodies
APC anti-human Galectin-9 [9M1-3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: ICFC, FC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Galectin-9
Clonality: Monoclonal
Clone: 9M1-3
Conjugation: APC (Allophycocyanin)
More Product Information
Order Now

SPECIFICATIONS

Antigen Name Galectin-9
Environmentally Preferable
Antigen Synonyms Tumor antigen HOM-HD-21|Lgals9|Ecalectin|GAL9|Gal-9
Antibody Type Primary
Clonality Monoclonal
Conjugation APC (Allophycocyanin)
Clone 9M1-3
Reactivity Baboon,African green monkey,Cynomolgus monkey,Human
Host Mouse
Gene ID 3965
Isotype IgG1 kappa
Flow Cytometry Yes
Immunogen Recombinant peptide from C-terminus of Galectin-9
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR