Autoclavable Single-Channel Pipettors, Labnet International
Supplier: Labnet International
P3941-10ISC-K P3941-200ISC-K P3941-100ISC-P P3941-20ISC-G P3941-20ISC-P P3941-SK4ISC-P
490014-652EA 722.27 USD
490014-652 490015-154 490014-642 490014-620 490014-634 490014-614
Autoclavable Single-Channel Pipettors, Labnet International
Pipettes Micropipettes
The pipettors have a new and improved design. Features include volume adjustment from dispenser button or thumbwheel, volume adjustment locking mechanism, and large volume display window.
  • Extended finger rest for greater pipetting comfort
  • Continuously adjust volume increments
  • Improved ergonomic design
  • Low dispensing and ejector forces
  • In-Lab calibration tool provided
  • Remodeled handle design for enhanced 'feel'
  • Now with even lower aspiration and dispense forces
  • Fully autoclavable
There are also extra ejection bar spacers provided for wide tip compatibility and chemically resistant nose cones. New tip ejector system on 2 to 1000 µl. Pipettors has Safe Click-to-Fix system, easy height adjustment, and fits virtually all brands of tips.
Ordering information: Three year warranty against manufacturing defects included. Supplied in four different colors.