Carbenicillin
Supplier: TEKNOVA INC
L8185
L8185
100219-764EA
115.67
USD
CA100219-764
100219-764
Carbenicillin
Carbenicillin
1.0% Tryptone, 0.5% yeast extract, 1.0% NaCl with 100 g/mL carbenicillin.Applications: Suitable for use with carbenicillin (ampicillin) resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC, etc.
Formula:
C₁₇H₁₈N₂O₆S |
CAS Number:
4697-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...