Tissue-Tek® Film® Coverslipper, Sakura Finetek
Supplier: Sakura
4743 4740
89199-930EA 104456.11 USD
89199-930 10204-846
Tissue-Tek® Film® Coverslipper, Sakura Finetek
Coverslippers
Coverslipper slides at a rate of 1080 per hour using flexible film
Slides are held in a carousel with a full capacity of 240 slides. Film is activated by xylene. Choice of 4 lengths of 45, 50, 55 or 60 mm.
Can be used as a stand alone instrument or linked to Sakura Prisma Stainer. The unit equipped with barcode reader offers ASCII data output that is easily read by computer systems with a USB interface. Most 2D and 1D barcode formats are supported.
Ordering information: Warranty is one year, parts and labor.