Hypersil GOLD™ CN HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25802-052130 25803-011001 25803-154630 25805-254630 25803-101030 25802-152130 25805-012106 25805-254030 25805-154630 25805-259270 25802-102130 25803-103030 25805-014001 25803-012101 25803-014001
10045-674EA 1390.45 USD
10045-674 10045-678 10045-714 10045-690 10045-680 10045-670 10041-690 10045-694 10045-684 10041-730 10041-742 10041-744 10045-706 10041-746 10045-718
Hypersil GOLD™ CN HPLC Columns, Thermo Scientific
Chromatography Columns
For alternative selectivity in both reversed phase and normal phase chromatography, use Thermo Scientific™ Hypersil GOLD™ CN HPLC columns.
Based on highly pure silica, these columns offer excellent peak shapes and outstanding sensitivity with less retention for faster analysis. Analyze a wide range of compounds, from steroids and polyphenols in reversed phase mode to surfactants and other polar species in normal phase mode.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).