Hypersil GOLD™ aQ HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25302-050365 25302-057581 25302-102130-V 25302-152130-V 25302-202130-V 25305-050165 25305-057581 25305-057582 25305-100165 25305-107582 25305-150165 25303-050265 25303-050365 25303-050565 25303-100365 25303-150265
10041-318EA 1071.98 USD
10041-318 10041-322 10041-326 10041-328 10041-330 10041-332 10041-334 10041-336 10041-338 10041-340 10041-342 10041-344 10041-346 10041-348 10041-370 10041-374 10041-376 10041-378 10041-380 10041-382 10041-392 10041-394 10041-398 10041-400 10041-410 10041-412 10041-414 10041-416 10041-420 10041-430 10041-436 10041-446 10045-502 10045-508 10045-510 10045-512 10045-516 10045-518 10045-520 10045-522 10045-524 10045-526 10045-528 10045-530 10045-532 10045-536 10045-538 10045-540 10045-542 10045-544 10045-546 10045-548 10045-550
Hypersil GOLD™ aQ HPLC Columns, Thermo Scientific
Chromatography Columns
Hypersil GOLD™ aQ is a polar endcapped C18 phase, which offers superior retention of polar compounds.
  • Ensures optimized peak shape, resolution, sensitivity and efficiency
  • Provides only controlled secondary interactions to ensure excellent peak shape for all analyte types
  • Polar endcapping of media also makes it usable in 100% aqueous mobile phases without the risk of loss of performance or poor stability
The additional interaction mechanism often provides selectivity differences over the traditional alkyl chain chemistries, and offers a solution forMore Product Information