Hypersil GOLD™ C8 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25202-100365 25205-100365 25205-109070 25205-109370 25205-254630 25203-100365 25205-259070A 25205-259270A 25205-018823A 25205-019023A 25205-259270XL
10041-198EA 1137.07 USD
10041-198 10041-200 10041-202 10041-206 10041-208 10041-210 10041-212 10041-214 10041-216 10041-220 10041-222 10041-226 10041-228 10041-232 10041-236 10041-240 10041-242 10041-246 10041-248 10041-252 10041-254 10041-256 10041-258 10041-262 10041-266 10041-270 10041-272 10041-276 10041-278 10041-280 10041-282 10041-284 10041-290 10041-292 10041-294 10041-296 10041-298 10041-300 10041-302 10041-304 10041-306 10041-308 10041-310 10041-312 10045-476 10045-480 10045-482 10045-484 10045-488 10045-490 10045-492 76332-204 76332-206 76332-208 76332-210 76332-372
Hypersil GOLD™ C8 HPLC Columns, Thermo Scientific
Chromatography Columns
For analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention, use Thermo Scientific™ Hypersil GOLD™ C8 HPLC columns for outstanding sensitivity and excellent peak shape.
The selectivity is similar to C18 columns with a predictable elution order but less retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).