PTFE Rubber Laminated Discs, Thermo Scientific
Supplier: Thermo Scientific
TS-12412 TS-12418 TS-12422
10055-946EA 108.18 USD
10055-946 10055-950 10055-970
PTFE Rubber Laminated Discs, Thermo Scientific
Septa
Provides a highy inert and unreactive seal.
  • Rigid and more difficult to puncture
  • Disc thickness of 60 mils
Made of white pharmaceutical rubber with 5 mils of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert nonreactive seal is desired. Discs are autoclavable with no loss of integrity after heating above 100 °C for five hours.